Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epithelial Specific Antigen Mouse anti-Human, Clone: VU-1D9, MilliporeSigma™

Catalog No. CBL251MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL251MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL251MI Supplier MilliporeSigma Supplier No. CBL251
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Epithelial Specific Antigen Antibody, clone VU-1D9 is a highly specific mouse monoclonal antibody, that targets Epithelial Cell Marker and has been tested in western blotting, immunocytochemistry and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Epithelial Specific Antigen
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone VU-1D9
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_002345
Gene Symbols EPCAM, GA733-2, M1S2, M4S1, MIC18, TACSTD1, TROP1
Host Species Mouse
Immunogen NCI-H69 whole cells
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.