Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epithelium/Endothelial Cells Mouse anti-Human, Clone: 18.29 (PHM-5), MilliporeSigma™

Catalog No. MAB430MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB430MI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB430MI Supplier MilliporeSigma Supplier No. MAB430

Mouse Monoclonal Antibody

Anti-Epithelium/Endothelial Cells Antibody, clone 18.29 (PHM-5) detects level of Epithelium/Endothelial Cells and has been published and validated for use in immunohistochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Epithelium/Endothelial Cells
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 18.29 (PHM-5)
Conjugate Unconjugated
Formulation Liquid in PBS, pH 7.4, with 0.2% BSA and 0.1% sodium azide.
Gene Accession No. NM_001018111.2; NM_005397.3
Gene Symbols PODXL, MGC138240, PCLP, podocalyxin-like, Gp200, PCLP1
Host Species Mouse
Quantity 500 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.