Learn More
EPM2AIP1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human EPM2AIP1 peptide using ARM Technology.
Supplier: Abnova Corporation H00009852K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The EPM2A gene, which encodes laforin, is mutated in an autosomal recessive form of adolescent progressive myoclonus epilepsy. The protein encoded by this gene binds to laforin, but its function is not known. This gene is intronless. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
EPM2AIP1 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
EPM2AIP1 | |
EPM2AIP1 | |
A synthetic peptide of human EPM2AIP1 | |
RUO | |
9852 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
EPM2A (laforin) interacting protein 1 | |
EPM2AIP1 | |
FLJ11207, KIAA0766 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.