Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPRS Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222311
Description
EPRS Polyclonal specifically detects EPRS in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| EPRS | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| bifunctional aminoacyl-tRNA synthetase, Cell proliferation-inducing gene 32 protein, DKFZp313B047, EARS, EC 6.1.1, EC 6.1.1.15, EC 6.1.1.18, GLNS, GLUPRORS, glutamate tRNA ligase, glutaminyl-tRNA synthetase, glutamyl-prolyl-tRNA synthetase, PARSproliferation-inducing gene 32 protein, proliferation-inducing protein 32, proline tRNA ligase, proline-tRNA ligase, prolyl-tRNA synthetase, QARS, QPRS | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 2058 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:10000, Immunohistochemistry 1:500 - 1:2000, Immunoprecipitation 2 - 10 ug/mg, Immunohistochemistry-Paraffin 1:500 - 1:2000 | |
| NP_004437.2 | |
| EPRS | |
| The immunogen this antibody was made to, maps to a region between residue 950 to 1000 of human Glutamyl-prolyl-tRNA Synthetase using the numbering given in entry NP_004437.2 (GeneID 2058). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction