Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eps8 Mouse, Unlabeled, Clone: 15, BD

Catalog No. BDB610144 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610144 150 μg
BDB610143 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610144 Supplier BD Biosciences Supplier No. 610144
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The p97 [Eps8] protein, a substrate for EGF-R tyrosine kinase, contains an SH3 domain, but lacks a functional SH2 domain. Antibodies to Eps8 recognize the 97 kDa protein and a less abundant 68 kDa protein. Both forms are tyrosine-phosphorylated following treatment of cells with EGF. It is likely that p68 [Eps8] is synthesized from an alternatively spliced mRNA since two major Eps8-specific mRNAs are detected by Northern analysis. Co-immunoprecipitation studies have demonstrated a physical association between the Eps8 protein and the EGF-R both in vivo and in vitro. For many EGF-R substrates, this interaction is mediated through an SH2 domain of the substrate. Since Eps8 lacks a well defined SH2 domain and a fusion protein containing the SH2-like region of Eps8 could not bind EGF-R, the mechanism of Eps8-EGF-R association remains unclear. Overexpression of Eps8 in fibroblasts and hematopoietic cells results in an increased mitogenic response to EGF, suggesting that Eps8 has a role in the modulation of EGF-R function.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Eps8
Applications Western Blot
Classification Monoclonal
Clone 15
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.