Learn More
Equilibrative Nucleoside Transporter 1 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Equilibrative Nucleoside Transporter 1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. |
| Gene Symbols | SLC29A1; ENT1 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human Equilibrative Nucleoside Transporter 1. Immunogen is 80% identical to mouse and rat ENT1. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.