Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

erbB-3/HER-3 Mouse anti-Human, Rat, Mouse, Bovine, Clone: 2F12, MilliporeSigma™

Catalog No. 05390MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-390-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-390-MI Supplier MilliporeSigma Supplier No. 05390
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect erbB-3/HER-3 using this Anti-erbB-3/HER-3 Antibody, clone 2F12 validated for use in immunocytochemistry, immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen erbB-3/HER-3
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2F12
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 10mM PBS, pH 7.4, 0.2% BSA, with 0.09% sodium azide. Frozen solution.
Gene Accession No. NM_001982
Gene Symbols ERBB3, c-erbB-3, c-erbB3, erbB-3, p180-ErbB3, p85-sErbB3, p45-sErbB3, ErbB-3, erbB3-S, MGC88033, HER3, MDA-BF-1
Host Species Mouse
Immunogen Recombinant rat erbB-3/HER-3 oncoprotein expressed in insect cells and purified by metal chelating chromatography. The epitope maps between amino acid residues 1295-1323 of human erbB-3. Clone 2F12.
Purification Method Protein A chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.