Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

erbB4 Mouse anti-Human, Mouse, Rat, Clone: HFR1/2G4, MilliporeSigma™

Catalog No. 051133MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-113-3MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-113-3MI Supplier MilliporeSigma Supplier No. 051133
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect erbB4 using this Anti-erbB4 Antibody, clone HFR1/2G4 validated for use in western blot, immunoprecipitation, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen erbB-4/HER-4
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone HFR1/2G4
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgM in 0.02MPB with 0.25MNaCl, pH 7.6, 0.01% Sodium Azide.
Gene Accession No. NP_005226.1
Gene Symbols ERBB4, c-erbB-4, c-erbB4, p180erbB4, erbB-4, MGC138404, HER4, EC 2.7.10
Host Species Mouse
Immunogen Synthetic peptide sequence RSTLQHPDYLQEYST from the cytoplasmic domain of the protein, residues 1250-1264.
Purification Method Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Protect from light.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.