Learn More
anti-ErbB3/Her3, PerCP, Clone: 66223, R&D Systems™
Supplier:  R&D Systems FAB3481C025
 View more versions of this product
Catalog No. FAB3481C025
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
ErbB3/Her3
Monoclonal
PerCP
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
2065
Human
Purified
Flow Cytometry
66223
Flow Cytometry 10 uL/10^6 cells
P21860
ERBB3
Mouse myeloma cell line NS0-derived recombinant human ErbB3/Her3, Ser20-Thr643, Accession # P21860
25 Tests
Primary
Detects human ErbB3/Her3 in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant human EGF R is observed.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.