Learn More
excision repair cross-complementing rodent repair deficiency, complementation group 2, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ERCC2 protein.
Supplier: Abnova Corporation H00002068B01P
Description
The nucleotide excision repair pathway is a mechanism to repair damage to DNA. The protein encoded by this gene is involved in transcription-coupled nucleotide excision repair and is an integral member of the basal transcription factor BTF2/TFIIH complex. The gene product has ATP-dependent DNA helicase activity and belongs to the RAD3/XPD subfamily of helicases. Defects in this gene can result in three different disorders, the cancer-prone syndrome xeroderma pigmentosum complementation group D, trichothiodystrophy, and Cockayne syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MKLNVDGLLVYFPYDYIYPEQFSYMRELKRTLDAKGHGVLEMPSGTGKTVSLLALIMAYQRAYPLEVTKLIYCSRTVPEIEKVIEELRKLLNFYEKQEGEKLPFLGLALSSRKNLCIHPEVTPLRFGKDVDGKCHSLTASYVRAQYQHDTSLPHCRFYEEFDAHGREVPLPAGIYNLDDLKALGRRQGWCPYFLARYSILHANVVVYSYHYLLDPKIADLVSKELARKAVVVFDEAHNIDNVCIDSMSVNLTRRTLDRCQGNLETLQKTVLRIKETDEQRLRDEYRRLVEGLREASAARETDAHLANPVLPDEVLQEAVPGSIRTAEHFLGFLRRLLEYVKWRLRVQHVVQESPPAFLSGLAQRVCIQRKPLRFCAERLRSLLHTLEITDLADFSPLTLLANFATLVSTYAKGFTIIIEPFDDRTPTIANPILHFSCMDASLAIKPVFERFQSVIITSGTLSPLDIYPKILDFHPVTMATFTMTLARVCLCPMIIGRGNDQVAISSKFETREDIAVIRNYGNLLLEMSAVVPDGIVAFFTSYQYMESTVASWYEQGILENIQRNKLLFIETQDGAETSVALEKYQEACENGRGAILLSVARGKVSEGIDFVHHYGRAVIMFGVPYVYTQSRILKARLEYLRDQFQIRENDFLTFDAMRHAAQCVGRAIRGKTDYGLMVFADKRFARGDKRGKLPRWIQEHLTDANLNLTVDEGVQVAKYFLRQMAQPFHREDQLGLSLLSLEQLESEETLKRIEQIAQQLSpecifications
excision repair cross-complementing rodent repair deficiency, complementation group 2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human ERCC2 protein. | |
ERCC2 | |
COFS2/EM9/MGC102762/MGC126218/MGC126219/TTD/XPD | |
Mouse | |
Affinity Purified | |
RUO | |
2068 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Immunofluorescence, Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_000400 | |
ERCC2 | |
ERCC2 (NP_000391.1, 1 a.a. ∼ 760 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.