Learn More
ERF, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human ERF peptide using ARM Technology.
Supplier: Abnova Corporation H00002077K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Members of the ETS family of transcription factors, such as ERF, regulate cell proliferation and differentiation. They share a highly conserved DNA-binding domain, the ETS domain, that recognizes the sequence GGAA/T (de Castro et al., 1997 [PubMed 9192842]). For further information on ETS transcription factors, see ETS1 (MIM 164720).[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
ERF | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
ERF | |
ERF | |
A synthetic peptide of human ERF | |
RUO | |
2077 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
Ets2 repressor factor | |
ERF | |
PE-2, PE2 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.