Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERK (pan ERK) Mouse, Unlabeled, Clone: 16, BD

Catalog No. BDB610123 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610123 50 μg
BDB610124 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610123 Supplier BD Biosciences Supplier No. 610123
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The family of serine/threonine kinases known as ERKs (extracellular signal regulated kinases) or MAPKs (mitogen-activated protein kinases) are activated after cell stimulation by a variety of hormones and growth factors. Cell stimulation induces a signaling cascade that leads to phosphorylation of MEK (MAPK/ERK kinase) which, in turn, activates ERK via tyrosine and threonine phosphorylation. A myriad of proteins represent the downstream effectors for the active ERK and implicate it in the control of cell proliferation and differentiation, as well as regulation of the cytoskeleton. Activation of ERK is normally transient and cells possess dual specificity phosphatases that are responsible for its down-regulation. Furthermore, multiple studies have shown that elevated ERK activity is associated with some cancers. ERK1 may be observable migrating at 44kDa and ERK2 at 42kDa in addition to a 54kDa ERK and a MAP kinase in the 90kDa range.

Host Species: Mouse
Clone: 16
Isotype: IgG2a
Species Reactivity: Mouse
Immunogen: Rat ERK2 aa. 219-358
Formula Weight [Chemical]: 42-85kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen ERK (pan ERK)
Applications Western Blot
Classification Monoclonal
Clone 16
Concentration 250μg/mL
Conjugate Unconjugated
Description Extracellular signal Regulated Kinases
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat ERK2 aa. 219-358
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.