Learn More
ERK2, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA128196
Description
The protein encoded by this gene is a member of the MAP kinase family. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. Upon activation, this kinase translocates to the nucleus of the stimulated cells, where it phosphorylates nuclear targets. Two alternatively spliced transcript variants encoding the same protein, but differing in the UTRs, have been reported for this gene. , ,
PA1-28196 detects ERK2 from human, mouse, rat samples.PA1-28196 has been successfully used in immunohistochemistry (paraffin tissue) applications.The PA1-28196 immunogen is: Synthetic peptide (unfortunately, the amino acid sequence is considered to be commercially sensitive) (Mouse) (C terminal).
Specifications
p42 MAP Kinase/ERK2/MAPK1 | |
Polyclonal | |
MAPK1 | |
ERK, ERK2, ERT1, MAPK2, P42MAPK, PRKM1, PRKM2, p38, p40, p41, p41mapk | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5594 | |
Store at 4°C, do not freeze | |
Purified |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
P28482, P63085, P63086 | |
MAPK1 | |
Synthetic peptide corresponding to the Mouse C terminal region. | |
7 mL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.