Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

erythroblast membrane-associated protein (Scianna blood group), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018806 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-806 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-806 Supplier Abnova Corporation Supplier No. H00114625D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human ERMAP protein.

The protein encoded by this gene is a cell surface transmembrane protein that may act as an erythroid cell receptor, possibly as a mediator of cell adhesion. Polymorphisms in this gene are responsible for the Scianna/Radin blood group system. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Sequence: MEMASSAGSWLSGCLIPLVFLRLSVHVSGHAGDAGKFHVALLGGTAELLCPLSLWPGTVPKEVRWLRSPFPQRSQAVHIFRDGKDQDEDLMPEYKGRTVLVRDAQEGSVTLQILDVRLEDQGSYRCLIQVGNLSKEDTVILQVAAPSVGSLSPSAVALAVILPVLVLLIMVCLCLIWKQRRAKEKLLYEHVTEVDNLLSDHAKEKGKLHKAVKKLRSELKLKRAAANSGWRRARLHFVAVTLDPDTAHPKLILSEDQRCVRLGDRRQPVPDNPQRFDFVVSILGSEYFTTGCHYWEVYVGDKTKWILGVCSESVSRKGKVTASPANGHWLLRQSRGNEYEALTSPQTSFRLKEPPRCVGIFLDYEAGVISFYNVTNKSHIFTFTHNFSGPLRPFFEPCLHDGGKNTAPLVICSELHKSEESIVPRPEGKGHANGDVSLKVNSSLLPPKAPELKDIILSLPPDLGPALQELKAPSF

Specifications

Antigen erythroblast membrane-associated protein (Scianna blood group)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human ERMAP protein.
Formulation PBS with no preservative; pH 7.4
Gene ERMAP
Gene Accession No. NM_001017922.1
Gene Alias MGC118810/MGC118811/MGC118812/MGC118813/PRO2801/RD/SC
Gene Symbols ERMAP
Host Species Rabbit
Immunogen ERMAP (NP_001017922.1, 1 a.a. ∼ 475 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114625
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.