Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

endoplasmic reticulum to nucleus signaling 1, Mouse, Clone: 4E1, Abnova™

Catalog No. 89000642 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-642 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-642 Supplier Abnova Corporation Supplier No. H00002081M05
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ERN1.

The protein encoded by this gene is the ER to nucleus signalling 1 protein, a human homologue of the yeast Ire1 gene product. This protein possesses intrinsic kinase activity and an endoribonuclease activity and it is important in altering gene expression as a response to endoplasmic reticulum-based stress signals. [provided by RefSeq

Sequence: EEVINLVDQTSENAPTTVSRDVEEKPAHAPARPEAPVDSMLKDMATIILSTFLLIGWVAFIITYPLSMHQQQQLQHQQFQKELEKIQLLQQQQQQLPFHP

Specifications

Antigen endoplasmic reticulum to nucleus signaling 1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4E1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ERN1.
Formulation PBS with no preservative; pH 7.4
Gene ERN1
Gene Accession No. AF059198
Gene Alias FLJ30999/IRE1/IRE1P/MGC163277/MGC163279
Gene Symbols ERN1
Host Species Mouse
Immunogen ERN1 (AAC25991, 401 a.a. ∼ 500 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 2081
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.