Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERR-alpha P2 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABE64
Description
Use Anti-ERR-alpha P2 Antibody (Rabbit Polyclonal Antibody) validated in western blot, immunocytochemistry to detect ERR-alpha P2 also known as Estrogen receptor-like 1, Estrogen-related receptor alpha.
Specifications
ERR-alpha P2 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Protein A purfied | |
Primary | |
Human, Mouse | |
Purified |
Immunocytochemistry, Western Blot | |
Unconjugated | |
ERR-alpha; ERR1; ERRa; ERRalpha; ESRL1; NR3B1 | |
KLH-conjugated linear peptide corresponding to human ERR-alpha. | |
100 μg | |
NP_004442 | |
2-8°C for 12 months | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction