Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERRFI1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216372
Description
ERRFI1 Polyclonal specifically detects ERRFI1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ERRFI1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
ERBB receptor feedback inhibitor 1, GENE-33, MIG6, MIG-6Mitogen-inducible gene 6 protein, RALTreceptor-associated late transducer | |
Rabbit | |
51 kDa | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
0.65mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
Q9UJM3 | |
ERRFI1 | |
Recombinant protein encompassing a sequence within the center region of human Mig-6. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
54206 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction