Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythroid Cells Mouse anti-Rat, APC, Clone: HIS49, BD

Catalog No. BDB562348 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562348 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562348 Supplier BD Biosciences Supplier No. 562348
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HIS49 antibody reacts with an antigen found on erythrocytes and erythroid progenitors, but not on granulocytes, monocytes, or lymphocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Erythroid Cells
Applications Flow Cytometry
Classification Monoclonal
Clone HIS49
Concentration 0.2mg/mL
Conjugate APC
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat bone marrow cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.