Learn More
Erythroid Cells Mouse anti-Rat, Unlabeled, Clone: HIS49, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 550961
Description
The HIS49 antibody reacts with an antigen found on erythrocytes and erythroid progenitors,1 but not on granulocytes, monocytes, or lymphocytes.
Host Species: Mouse
Clone: HIS49
Isotype: IgM, κ
Species Reactivity: Rat
Immunogen: Rat bone marrow cells
Flow Cytometry, Immunohistochemistry (Frozen)

Specifications
Erythroid Cells | |
Monoclonal | |
0.5mg/mL | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
IgM |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen) | |
HIS49 | |
Unconjugated | |
Rat bone marrow cells | |
0.1 mg | |
Immunology | |
Rat |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.