Learn More
Description
Host Species: Rat
Clone: TER-119
Isotype: IgG2b, κ
Species Reactivity: Ms
Immunogen: Mouse Fetal Liver
Flow cytometry
Specifications
Specifications
| Antigen | Erythroid Cells |
| Classification | Monoclonal |
| Clone | TER-119 |
| Concentration | 0.2mg/mL |
| Conjugate | Brilliant Violet 421 |
| Host Species | Rat |
| Immunogen | Mouse Fetal Liver |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.