Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythropoietin Mouse anti-Human, Clone: 1C8, MilliporeSigma™

Catalog No. MAB1072MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1072MI 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1072MI Supplier MilliporeSigma Supplier No. MAB1072
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Erythropoietin using this Anti-Erythropoietin Antibody, clone 1C8 validated for use in ELISA, immunofluorescence and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Erythropoietin
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1C8
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2b in buffer containing 20mM sodium phosphate, 300mM NaCl, and 0.02% sodium merthiolate.
Gene Accession No. NM_000799.2
Gene Symbols EPO, MGC138142, EP
Host Species Mouse
Immunogen Recombinant human Epo.
Purification Method Protein A purfied
Quantity 250 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.