Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Erythropoietin R Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP119388
Description
Erythropoietin R Polyclonal specifically detects Erythropoietin R in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Erythropoietin R | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
EPO-R, erythropoietin receptor, MGC138358 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
EpoR (A364) detects endogenous levels of EpoR protein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Western Blot 2.0 ug/ml, Simple Western 1:10, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
P19235 | |
EPOR | |
A synthetic peptide made to an internal portion of the human EPO receptor protein (between residues 300-400) [UniProt P19235] | |
0.1 mL | |
Angiogenesis, Immunology, Signal Transduction | |
2057 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction