Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Estrogen Receptor α Rabbit anti-Human, Mouse, Rat, Bovine, Pig, Clone: 60C, MilliporeSigma™

Catalog No. 04820MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-820-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-820-MI Supplier MilliporeSigma Supplier No. 04820
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Anti-Estrogen Receptor α Antibody, clone 60C is an antibody against Estrogen Receptor α for use in western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Estrogen Receptor α
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 60C
Conjugate Unconjugated
Formulation Cultured supernantant with 0.05% sodium azide.
Gene Accession No. NP_000116.2
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 13-32 (C-ALLHQIQGNELEPLNRPQLK) of human Estrogen Receptor α. The immunizing sequence is identical in rat and mouse and has 19/20 identical amino acids in bovine and porcine.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) ER, ER-alpha, ESR, ESRA, Era, NR3A1, DKFZp686N23123, OTTHUMP00000017718, OTTHUMP00000017719
Target Species Bovine, Human, Mouse, Pig, Rat
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.