Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Estrogen Receptor Β Mouse anti-Rat, Mouse, Human, Clone: 9.88, MilliporeSigma™

Catalog No. MABE290MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE290MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE290MI Supplier MilliporeSigma Supplier No. MABE290
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-Estrogen Receptor β Antibody, clone 9.88 (Mouse Monoclonal Antibody) validated in western blot, immunoprecipitation to detect Estrogen Receptor β also known as Estrogen receptor beta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Estrogen Receptor β
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 9.88
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_001035365
Gene Symbols ESR2, ESTRB, NR3A2
Host Species Mouse
Immunogen Recombinant protein corresponding to human Estrogen Receptor β.
Purification Method Purified by ion-exchange chromatography.
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.