Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Estrogen Receptor β, Polyclonal,

Catalog No. 06629MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-629-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-629-MI Supplier MilliporeSigma™ Supplier No. 06629

Specifications

Antigen Estrogen Receptor β
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation UnPurified mouse ascites containing no preservatives.
Gene Symbols ESR2; ESR-BETA; ESTRB; ESRB; ER-BETA; NR3A2; Erb; ER-beta
Host Species Rabbit
Immunogen peptide (YAEPQKSPWCEARSLEHT) representing amino acids 54-71 of rat and mouse estrogen receptor β and amino acids 46-63 of human estrogen receptor β
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001437
Target Species Human, Primate, Rat
Content And Storage −20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.