Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Estrogen-related Receptor α Rabbit anti-Human, Hamster, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 07662MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-662-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-662-MI Supplier MilliporeSigma Supplier No. 07662
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Estrogen-related Receptor α Antibody is a Rabbit Polyclonal Antibody for detection of Estrogen-related Receptor α also known as ERR alpha and has been tested in EMSA, immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Estrogen-related Receptor α
Applications Electromobility Shift Assay, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Protein A Purified rabbit IgG in a solution of 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.135M NaCl, and 0.035% sodium azide as a preservative.
Gene Symbols ESR1; ER-alpha; ESR; Era; ER; NR3A1; ESRA; DKFZp686N23123
Host Species Rabbit
Immunogen Ovalbumin-conjugated, synthetic peptide corresponding to amino acids 339-364 of human Estrogen Receptor-Related α
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000125.2
Target Species Hamster, Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.