Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETFA, Mouse, Clone: 2A5, Abnova™

Catalog No. 89022011 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-011 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-011 Supplier Abnova Corporation Supplier No. H00002108M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant ETFA.

ETFA participates in catalyzing the initial step of the mitochondrial fatty acid beta-oxidation. It shuttles electrons between primary flavoprotein dehydrogenases and the membrane-bound electron transfer flavoprotein ubiquinone oxidoreductase. Defects in electron-transfer-flavoprotein have been implicated in type II glutaricaciduria in which multiple acyl-CoA dehydrogenase deficiencies result in large excretion of glutaric, lactic, ethylmalonic, butyric, isobutyric, 2-methyl-butyric, and isovaleric acids. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MFRAAAPGQLRRAASLLRFQSTLVIAEHANDSLAPITLNTITAATRLGGEVSCLVAGTKCDKVAQDLCKVAGIAKVLVAQHDVYKGLLPEELTPLILATQKQFNYTHICAGASAFGKNLLPRVAAKLEVAPISDIIAIKSPDTFVRTIYAGNALCTVKCDEKVKVFSVRGTSFDAAATSGGSASSEKASSTSPVEISEWLDQKLTKSDRPELTGAKVVVSGGRGLKSGENFKLLYDLADQLHAAVGASRAAVDAGFVPNDMQVGQTGKIVAPELYIAVGISGAIQHLAGMKDSKTIVAINKDPEAPIFQVADYGIVADLFKVVPEMTEILKKK

Specifications

Antigen ETFA
Applications ELISA
Classification Monoclonal
Clone 2A5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant ETFA.
Formulation PBS with no preservative; pH 7.4
Gene ETFA
Gene Accession No. BC015526
Gene Alias EMA/GA2/MADD
Gene Symbols ETFA
Host Species Mouse
Immunogen ETFA (AAH15526, 1 a.a. ∼ 333 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2108
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.