Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETS1 DBD Mouse anti-Human, Mouse, Rat, Clone: 1ET 7F2, MilliporeSigma™

Catalog No. 041542MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-154-2MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-154-2MI Supplier MilliporeSigma Supplier No. 041542
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-ETS1 DBD Antibody, clone 1ET 7F2 (Mouse Monoclonal Antibody) validated in western blotting to detect ETS1 DBD also known as Avian erythroblastosis virus E26 (v-ets) oncogene homolog-1, ets protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETS1 DBD
Applications Western Blot
Classification Monoclonal
Clone 1ET 7F2
Conjugate Unconjugated
Formulation Unpurified mouse monoclonal IgG1κ in preservative-free ascites.
Gene Accession No. NP_005229.1
Gene Symbols ETS1, ETS-1, p54, ETS-1 protein: ETS 1: ETS1, EWSR 2, EWSR2
Host Species Mouse
Immunogen Linear peptide corresponding to human ETS1 DBD.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.