Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EVX1, Mouse, Clone: 1B7, Abnova™

Catalog No. 89022017 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-017 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-017 Supplier Abnova Corporation Supplier No. H00002128M07
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant EVX1.

This gene encodes a member of the even-skipped homeobox family characterized by the presence of a homeodomain closely related to the Drosophila even-skipped (eve) segmentation gene of the pair-rule class. The encoded protein may play an important role as a transcriptional repressor during embryogenesis. [provided by RefSeq

Sequence: ESRKDMVVFLDGGQLGTLVGKRVSNLSEAVGSPLPEPPEKMVPRGCLSPRAVPPATRERGGGGPEEEPVDGLAGSAAGPGAEPQVAGAAMLGPGPPAPSVDSLSGQGQP*

Specifications

Antigen EVX1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1B7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant EVX1.
Formulation PBS with no preservative; pH 7.4
Gene EVX1
Gene Accession No. NM_001989
Gene Symbols EVX1
Host Species Mouse
Immunogen EVX1 (NP_001980, 2 a.a. ∼ 110 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2128
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.