Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Excitatory amino acid transporter 2 Mouse anti-Human, Mouse, Rat, Clone: G6, MilliporeSigma™

Catalog No. MAB2262MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2262MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2262MI Supplier MilliporeSigma Supplier No. MAB2262
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Excitatory amino acid transporter 2 Antibody, clone G6 is validated for use in western blot, immunohistochemistry, IC for the detection of Excitatory amino acid transporter 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Excitatory amino acid transporter 2
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone G6
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_004162
Gene Symbols SLC1A2, EAAT2, GLT-1, GLT1
Host Species Mouse
Immunogen Linear peptide corresponding to the C-terminus of human Excitatory amino acid transporter 2.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.