Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Excitatory Amino Acid Transporter (GLT-1), Polyclonal
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ PC154100UL
Description
Specifically detects Excitatory Amino Acid Transporter (GLT-1) Clone: in Rat samples, and it is validated for Immunoprecipitation
Precise regulation of excitatory amino acids (EAA) such as glutamate is essential, as glutamate is not only the major excitatory neurotransmitter, but also a potent excitotoxin. Defects in functional glutamate transport have been identified in the amyotrophic lateral sclerosis (ALS) brain and spinal cord, suggesting that defective glutamate transport may contribute to motor neuron degeneration. Three glutamate transporter subtypes have been identified in mammals, which include GLAST, GLT-1 and EAAC1. More recently, the homologous proteins EAAT1, EAAT2, and EAAT3 have been characterized in humans, respectively. EAAT4 and EAAT5 transporters have been localized to Purkinje cell synapses, and retina, respectively. The selective loss of glutamate transport in ALS appears to be specific for the GLT-1 (EAAT2) transporter subtyp. While GLT-1, GLAST and EAAC1 share approximately 50% sequence homology, the sequence used as the immunogen is unique to GLT-1.
Specifications
| Excitatory Amino Acid Transporter (GLT-1) | |
| Polyclonal | |
| Serum diluted in PBS. | |
| a synthetic peptide (NGKSADCSVEEEPWKREK) corresponding to amino acids at the C-terminus of rat GLT-1 | |
| RUO | |
| Recognizes the ∽75kDa and the ∽50kDa forms of GLT-1. | |
| −20°C | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Rat | |
| Diluted |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction