Learn More
exostoses (multiple)-like 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant EXTL1.
Supplier: Abnova Corporation H00002134A01
Description
This gene is a member of the multiple exostoses (EXT) family of glycosyltransferases, which function in the chain polymerization of heparan sulfate and heparin. The encoded protein harbors alpha 1,4- N-acetylglucosaminyltransferase activity, and is involved in chain elongation of heparan sulfate and possibly heparin. [provided by RefSeq
Sequence: AQTGECSSMPLQWNRGRNHLVLRLHPAPCPRTFQLGQAMVAEASPTVDSFRPGFDVALPFLPEAHPLRGGAPGQLRQHSPQPGVALLALEEERGGWRTADTSpecifications
exostoses (multiple)-like 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant EXTL1. | |
EXTL1 | |
EXTL/MGC70794 | |
Mouse | |
50 μL | |
Yes | |
2134 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_004455 | |
EXTL1 | |
EXTL1 (NP_004446, 141 a.a. ∼ 241 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.