Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EXTL2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526241
Description
EXTL2 is a glycosyltransferase required for the biosynthesis of heparan-sulfate and responsible for the alternating addition of beta-1-4-linked glucuronic acid (GlcA) and alpha-1-4-linked N-acetylglucosamine (GlcNAc) units to nascent heparan sulfate chains.
Specifications
| EXTL2 | |
| Polyclonal | |
| EXTL2 | |
| EXTR2, EXT-related protein 2, alpha-1,4-N-acetylhexosaminyltransferase EXTL2, alpha-1,4-N-acteylhexosaminyltransferase, alpha-GalNAcT EXTL2, exostosin-like 2, glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase, processed exostosin-like 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2135, 58193 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Q9UBQ6, Q9ES89 | |
| EXTL2 | |
| KLH conjugated synthetic peptide between 269-295 amino acids from the C-terminal region of human EXTL2 | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction