Learn More
Description
This gene encodes a member of the eyes absent (EYA) family of proteins. The encoded protein may be post-translationally modified and may play a role in eye development. A similar protein in mice can act as a transcriptional activator. Alternative splicing results in multiple transcript variants, but the full-length natures of all of these variants have not yet been determined. [provided by RefSeq
Specifications
Specifications
| Antigen | eyes absent homolog 2 (Drosophila) |
| Applications | ELISA, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 2F8 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant EYA2. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | EYA2 |
| Gene Accession No. | NM_172110 |
| Gene Alias | EAB1/MGC10614 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.