Learn More
eyes absent homolog 2 (Drosophila), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human EYA2 protein.
Supplier: Abnova Corporation H00002139D01P
Description
This gene encodes a member of the eyes absent (EYA) family of proteins. The encoded protein may be post-translationally modified and may play a role in eye development. A similar protein in mice can act as a transcriptional activator. Alternative splicing results in multiple transcript variants, but the full-length natures of all of these variants have not yet been determined. [provided by RefSeq
Sequence: MVELVISPSLTVNSDCLDKLKFNRADAAVWTLSDRQGITKSAPLRVSQLFSRSCPRVLPRQPSTAMAAYGQTQYSAGIQQATPYTAYPPPAQAYGIPSYSIKTEDSLNHSPGQSGFLSYGSSFSTSPTGQSPYTYQMHGTTGFYQGGNGLGNAAGFGSVHQDYPSYPGFPQSQYPQYYGSSYNPPYVPASSICPSPLSTSTYVLQEASHNVPNQSSESLAGEYNTHNGPSTPAKEGDTDRPHRASDGKLRGRSKRSSDPSPAGDNEIERVFVWDLDETIIIFHSLLTGTFASRYGKDTTTSVRIGLMMEEMIFNLADTHLFFNDLEDCDQIHVDDVSSDDNGQDLSTYNFSADGFHSSAPGANLCLGSGVHGGVDWMRKLAFRYRRVKEMYNTYKNNVGGLIGTPKRETWLQLRAELEALTDLWLTHSLKALNLINSRPNCVNVLVTTTQLIPALAKVLLYGLGSVFPIENIYSATKTGKESCFERIMQRFGRKAVYVVIGDGVEEEQGAKKHNMPFWRISCHADLEALRHALELEYLSpecifications
eyes absent homolog 2 (Drosophila) | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human EYA2 protein. | |
EYA2 | |
EAB1/MGC10614 | |
Rabbit | |
Affinity Purified | |
RUO | |
2139 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_005244.3 | |
EYA2 | |
EYA2 (NP_005235.3, 1 a.a. ∼ 538 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.