Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EZH2 Mouse, Unlabeled, Clone: 11, BD

Catalog No. BDB612667 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612667 150 μg
BDB612666 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612667 Supplier BD Biosciences Supplier No. 612667
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 11/EZH2 monoclonal antibody specifically binds to the methyltransferase, EZH2 (Enhancer of Zeste Homolog 2). EZH2 is a human homologue of Drosophila's Enhancer of zeste gene, an important regulator of homeobox gene expression. The EZH2 protein has a predicted molecular weight of ∼85 kDa. EZH2 is a member of the Polycomb group (PcG) of proteins that are essential for the maintenance, but not initiation, of the transcriptionally repressed state of certain developmental genes. PcG proteins are a structurally diverse group of proteins with conserved functions from fly to human cells. PcG family proteins form multimeric complexes that regulate the expression of genes involved in cell cycle, DNA repair and differentiation. Specifically, EZH2 is a core enzymatic component of PRC2 (polycomb repressive complex 2). EZH2 is expressed in some lymph node follicular T cells and B cells. Thymocytes differentially express EZH2 at various stages during T-cell maturation. EZH2 interacts with multiple signaling proteins, including Vav, that are involved in lymphocyte development and activation. It is highly expressed in a variety of tumors including lymphomas as well as breast and prostate cancers. EZH2 is important in the self renewal and proliferation of numerous stem cell types including fetal hematopoietic stem cells, muscle satellite cells, hepatic stem/progenitor cells, neural stem cells, basal cell progenitors in the developing epidermis, embryonic stem cells, and some cancer stem cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EZH2
Applications Western Blot
Classification Monoclonal
Clone 11
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human EZH2 recombinant protein aa. 156-256
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.