Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EZH2 Mouse anti-Human, Mouse, Clone: BD43, MilliporeSigma™

Catalog No. 051319MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-131-9MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-131-9MI Supplier MilliporeSigma Supplier No. 051319
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-EZH2 Antibody, clone BD43 (Mouse Monoclonal Antibody) validated in western blotting to detect EZH2 also known as Lysine N-methyltransferase 6, enhancer of zeste homolog 2, ENX-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EZH2
Applications Western Blot
Classification Monoclonal
Clone BD43
Conjugate Unconjugated
Formulation Mouse monoclonal IgG1κ ascites in buffer containing 0.1MTris-Glycine (pH7.4) 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_004447.2
Gene Symbols EZH2, KMT6
Host Species Mouse
Immunogen Recombinant fusion protein of amino acids 1-286 of human EZH2 tagged with MBP.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.