Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ezrin (pT567) Mouse, Unlabeled, Clone: J37-954.281.307, BD

Catalog No. BDB558357 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558357 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558357 Supplier BD Biosciences Supplier No. 558357

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Ezrin is a member of the ERM (Ezrin-Radixin-Moesin) family of proteins that can function as a crosslinker between the actin cytoskeleton and the plasma membrane of the cell. Phosphorylation of threonine residues (Thr567 on Ezrin, Thr564 on Radixin and Thr558 on Moesin) has been reported to occur with stimulation of growth factors and is important for cytoskeletal rearrangements. Phosphorylation of Thr567 at the C-terminal F actin-binding domain has been reported to activate the conversion of Ezrin from a dormant soluble form in the cytosol to a membrane- and actin-binding conformation. In addition to its reactivity to the human species, the peptide used to generate this antibody is either identical or has homology to analogous regions of mouse and rat Ezrin, Radixin and Moesin. The J37-954.281.307 monoclonal antibody has been reported to recognize phosphorylated T567 on active Ezrin in addition to crossreacting on phosphorylated T564 on Radixin and phosphorylated T558 on Moesin.

Immunohistochemistry, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ezrin (pT567)
Applications Western Blot
Classification Monoclonal
Clone J37-954.281.307
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Ezrin Peptide
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.