Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ezrin (pY353) Mouse anti-Human, Alexa Fluor 488, Clone: I66-386, BD

Catalog No. BDB558627 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558627 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558627 Supplier BD Biosciences Supplier No. 558627
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ezrin is a member of the ERM (ezrin-radixin-moesin) family of proteins that function as crosslinkers between the actin cytoskeleton and the plasma membrane. Ezrin is a substrate for the tyrosine kinase epidermal (EGF) and hepatocyte growth factor (HGF) receptors. Upon EGF or HGF stimulation, ezrin is phosphorylated on tyrosine 353 (Y353), which is required for its interaction with phosphatidylinositol 3 kinase p85 subunit and activation of Akt. The I66-386 monoclonal antibody recognizes the phosphorylated Y353 of human ezrin.

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ezrin (pY353)
Applications Flow Cytometry
Classification Monoclonal
Clone I66-386
Conjugate Alexa Fluor 488
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human Ezrin Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.