Learn More
Ezrin/Radixin/Moesin Rabbit anti-Human, Mouse, Monkey, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Ezrin/Radixin/Moesin |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Protein A and peptide affinity purified immunoglobulin. Liquid in 10mM sodium HEPES, pH 7.5, 150mM NaCl, 100μg/mL BSA, with 50% glycerol. |
| Gene Symbols | SLC9A3R1; +exchanger); NHERF1; +H; NHERF; Na; NHERF-1; EBP50 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated, synthetic peptide corresponding to residues surrounding Thr567 of human ezrin. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.