Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ezrin (Thr567)/Radixin (Thr564)/Moesin (Thr558) Rabbit anti-Human, Mouse, Monkey, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB3832MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3832MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3832MI Supplier MilliporeSigma Supplier No. AB3832
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Ezrin (Thr567)/Radixin (Thr564)/Moesin (Thr558) Antibody, phospho-specific detects level of Ezrin (Thr567)/Radixin (Thr564)/Moesin (Thr558) and has been published and validated for use in immunohistochemistry (paraffin) and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ezrin (Thr567)/Radixin (Thr564)/Moesin (Thr558)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid in 10mM sodium HEPES, pH 7.5, 150mM NaCl, 100μg/mL BSA, with 50% glycerol.
Gene Symbols SLC9A3R1; +exchanger); NHERF1; +H; NHERF; Na; NHERF-1; EBP50
Host Species Rabbit
Immunogen KLH-conjugated, synthetic phospho-peptide corresponding to residues surrounding Thr567 of human ezrin
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_004252.2
Target Species Human, Monkey, Mouse, Rat
Content And Storage −20°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.