Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F11 Receptor Mouse anti-Human, Unlabeled, Clone: M.AB.F11, BD

Catalog No. BDB552147 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552147 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552147 Supplier BD Biosciences Supplier No. 552147

Mouse Monoclonal Antibody

Reacts with the F11 receptor (also known as JAM-1), an integral membrane protein duplex of 32-35kDa expressed on platelets, leukocytes, red blood cells, endothelial cells, epithelial cells, and various cell lines. The F11R is homologous with the murine protein called Junctional Adhesion Molecule (JAM), a member of the immunoglobulin superfamily. Monoclonal antibody (mAb) F11 is a stimulatory antibody, namely, the binding of F11 mAb to F11R induces morphological changes, granular secretion, and aggregation in human platelets that are mediated by two pathways. Antibody-induced platelet aggregation and granule secretion are dependent on both molecules of FcγRII and GPIIb/IIIa integrin, and that may play a role in some thrombocytopenias. F11R-mediated platelet adhesion and aggregation are not dependent on FcγRII or GPIIb/IIIa and that may play a role in normal platelet physiological processes.

Host Species: Mouse
Clone: M.Ab.F11
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen F11 Receptor
Applications Flow Cytometry
Classification Monoclonal
Clone M.AB.F11
Concentration 0.5mg/mL
Conjugate Unconjugated
Description JAM-1; Platelet F11 receptor; F11R; JAM-A; PAM-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.