Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

coagulation factor II (thrombin) receptor-like 3, Mouse, Clone: 5F4, Abnova™

Catalog No. 89014232 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-232 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-232 Supplier Abnova Corporation Supplier No. H00009002M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant F2RL3.

Coagulation factor II (thrombin) receptor-like 3 (F2RL3) is a member of the large family of 7-transmembrane-region receptors that couple to guanosine-nucleotide-binding proteins. F2RL3 is also a member of the protease-activated receptor family. F2RL3 is activated by proteolytic cleavage of its extracellular amino terminus. The new amino terminus functions as a tethered ligand and activates the receptor. F2RL3 is activated by thrombin and trypsin. [provided by RefSeq

Sequence: GGTQTPSVYDESGSTGGGDDSTPSILPAPRGYPGQVCANDSDTLELPDSSRALLLGWVPTRLVPA

Specifications

Antigen coagulation factor II (thrombin) receptor-like 3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5F4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant F2RL3.
Formulation PBS with no preservative; pH 7.4
Gene F2RL3
Gene Accession No. NM_003950
Gene Alias PAR4
Gene Symbols F2RL3
Host Species Mouse
Immunogen F2RL3 (NP_003941, 18 a.a. ∼ 82 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Membrane Receptors
Primary or Secondary Primary
Gene ID (Entrez) 9002
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.