Learn More
coagulation factor III (thromboplastin, tissue factor), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human F3 protein.
Supplier: Abnova Corporation H00002152D01P
Description
This gene encodes coagulation factor III which is a cell surface glycoprotein. This factor enables cells to initiate the blood coagulation cascades, and it functions as the high-affinity receptor for the coagulation factor VII. The resulting complex provides a catalytic event that is responsible for initiation of the coagulation protease cascades by specific limited proteolysis. Unlike the other cofactors of these protease cascades, which circulate as nonfunctional precursors, this factor is a potent initiator that is fully functional when expressed on cell surfaces. There are 3 distinct domains of this factor: extracellular, transmembrane, and cytoplasmic. This protein is the only one in the coagulation pathway for which a congenital deficiency has not been described. [provided by RefSeq
Sequence: METPAWPRVPRPETAVARTLLLGWVFAQVAGASGTTNTVAAYNLTWKSTNFKTILEWEPKPVNQVYTVQISTKSGDWKSKCFYTTDTECDLTDEIVKDVKQTYLARVFSYPAGNVESTGSAGEPLYENSPEFTPYLETNLGQPTIQSFEQVGTKVNVTVEDERTLVRRNNTFLSLRDVFGKDLIYTLYYWKSSSSGKKTAKTNTNEFLIDVDKGENYCFSVQAVIPSRTVNRKSTDSPVECMGQEKGEFREIFYIIGAVVFVVIILVIILAISLHKCRKAGVGQSWKENSPLNVSSpecifications
coagulation factor III (thromboplastin, tissue factor) | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human F3 protein. | |
F3 | |
CD142/TF/TFA | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse |
Western Blot | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
NM_001993 | |
F3 | |
F3 (AAH11029.1, 1 a.a. ∼ 295 a.a) full-length human protein. | |
100 μg | |
Membrane Receptors | |
2152 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.