Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-F4/80 FITC, Clone: BM8.1,
Rat Monoclonal Antibody
Supplier: MilliporeSigma™ MABF1527
Description
Specifically detects F4/80 Clone: BM8.1 in Mouse samples, and it is validated for Flow Cytometry
The BM8.1 antibody is specific for mouse F4/80 antigen, a 125kDa transmembrane protein widely expressed by members of the mononuclear phagocyte system and considered to be a key marker for mature macrophage cells. F4/80 is differentially expressed during myeloid cell development, and may be regulated by certain cytokines within the tissue microenvironment. Other cell types shown to express this antigen include Langerhans cells, Kupffer cells and dendritic cell subsets. BM8.1 is widely used together with antibodies to CD115 (c-fms), CD11b and CD11c to identify myeloid / macrophage cells by flow cytometry.
Specifications
F4/80 | |
Monoclonal | |
FITC | |
Q61549 | |
Rat | |
Purified by size-exclusion chromatography | |
RUO | |
Primary | |
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze |
Flow Cytometry | |
BM8.1 | |
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
Emr1, Gpf480 | |
Proprietary immunogen | |
25 μg | |
Inflammation, Immunology | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction