Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F4/80-Like Receptor Rat anti-Mouse, Unlabeled, Clone: 6F12, BD

Catalog No. BDB552958 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552958 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552958 Supplier BD Biosciences Supplier No. 552958
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 6F12 antibody reacts with a 7-transmembrane-domain protein, which is similar to the F4/80 macrophage antigen of the EGF-TM7 protein family and is encoded by the Emr4 gene. The FIRE protein is expressed on myeloid cells with a denditic cell (DC) developmental potential, including subsets of DC and macrophages in the spleen and lymph nodes, most resident peritoneal macrophages, many peripheral blood monocytes, and a subpopulation of bone-marrow myeloid-cell progenitors. The protein is not detected on peripheral T and B lymphocytes, and it is down-regulated on thioglycollate-elicited peritoneal macrophages and on dendritic cells activated by GM-CSF, IFN-γ, anti-CD40, and LPS. Using soluble biotinylated fusion protein, a FIRE ligand was detected on a mouse IgG+ B lymphoma cell line (A20), but not on myeloid, fibroblast, or T-cell lines, suggesting that the FIRE protein may be involved in immunoregulatory interactions between antigen-presenting cells and B lymphocytes.

Host Species: Rat
Clone: 6F12
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: CHO cells expressing recombinant FIRE fusion protein

Flow Cytometry, Immunohistochemistry (Frozen)

TRUSTED_SUSTAINABILITY

Specifications

Antigen F4/80-Like Receptor
Applications Flow Cytometry
Classification Monoclonal
Clone 6F12
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Fire; Emr4; EGF-like module receptor 4; D17Ertd479e; Egf-tm7
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen CHO cells expressing recombinant FIRE fusion protein
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.