Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FA2H, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89209599 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-209-599 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-209-599 Supplier Abnova Corporation Supplier No. H00079152K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human FA2H peptide using ARM Technology.

Sphingolipids are a large class of lipids found in all eukaryotic cells and are involved in numerous cellular processes. The structural diversity of sphingolipids stems from more than 300 distinct head groups, as well as from modifications of the hydrophobic ceramide moiety. FA2H catalyzes a common modification of the ceramide moiety: hydroxylation at the 2 position of the N-acyl chain. Sphingolipids containing 2-hydroxy fatty acid are common in nervous and epidermal tissue. Glycosphingolipids containing a high proportion of 2-hydroxy fatty acid are critical components of myelin, and several very long chain ceramides with 2-hydroxy fatty acids are important for the permeability barrier function of epidermis (Alderson et al., 2004 [PubMed 15337768]).[supplied by OMIM

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen FA2H
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description fatty acid 2-hydroxylase
Formulation In 1X PBS, pH 7.4
Gene FA2H
Gene Accession No. FA2H
Gene Alias FAAH, FAH1, FAXDC1, FLJ25287, SCS7
Gene Symbols FA2H
Host Species Rabbit
Immunogen A synthetic peptide of human FA2H
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79152
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.