Learn More
Description
This gene encodes a protein that is responsible for the hydrolysis of a number of primary and secondary fatty acid amides, including the neuromodulatory compounds anandamide and oleamide. [provided by RefSeq
Specifications
Specifications
| Antigen | FAAH |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2G8 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant FAAH. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | FAAH |
| Gene Accession No. | NM_001441 |
| Gene Alias | FAAH-1/MGC102823/MGC138146 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.