Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAAH, Mouse, Clone: 4H8, Abnova™

Catalog No. 89022034 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-034 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-034 Supplier Abnova Corporation Supplier No. H00002166M07
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant FAAH.

This gene encodes a protein that is responsible for the hydrolysis of a number of primary and secondary fatty acid amides, including the neuromodulatory compounds anandamide and oleamide. [provided by RefSeq

Sequence: DLNAPGRATGAVSYTMLYNCLDFPAGVVPVTTVTAEDEAQMEHYRGYFGDIWDKMLQKGMKKSVGLPVAVQCVALPWQEELCLRFMREVERLMTPEKQSS

Specifications

Antigen FAAH
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4H8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant FAAH.
Formulation PBS with no preservative; pH 7.4
Gene FAAH
Gene Accession No. NM_001441
Gene Alias FAAH-1/MGC102823/MGC138146
Gene Symbols FAAH
Host Species Mouse
Immunogen FAAH (NP_001432, 480 a.a. ∼ 579 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2166
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.