Learn More
FAAH2, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human FAAH2 peptide using ARM Technology.
Supplier: Abnova Corporation H00158584K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a fatty acid amide hydrolase that shares a conserved protein motif with the amidase signature family of enzymes. The encoded enzyme is able to catalyze the hydrolysis of a broad range of bioactive lipids, including those from the three main classes of fatty acid amides; N-acylethanolamines, fatty acid primary amides and N-acyl amino acids. This enzyme has a preference for monounsaturated acyl chains as a substrate
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
FAAH2 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
FAAH2 | |
FAAH2 | |
A synthetic peptide of human FAAH2 | |
RUO | |
158584 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
fatty acid amide hydrolase 2 | |
FAAH2 | |
AMDD, FAAH-2, FLJ31204, RP11-479E16 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.